EMAIL THIS PAGE TO A FRIEND
GF88732693

Holmium

wire reel, 50mm, diameter 0.5mm, as drawn, 99.9%

Synonym: HO005105, Holmium

  • CAS Number 7440-60-0

  • Empirical Formula (Hill Notation) Ho

  • Molecular Weight 164.93

  •  MDL number MFCD00011049

  •  PubChem Substance ID 329813362

  •  NACRES NA.23

Purchase

Properties

assay   ≥99.9%
form   wire
mfr. no.   Goodfellow, 887-326-93
density   8.8 g/mL at 25 °C (lit.)
SMILES string   [Ho]
InChI   1S/Ho
InChI key   KJZYNXUDTRRSPN-UHFFFAOYSA-N

Description

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Safety & Documentation

Safety Information

WGK Germany 
nwg
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?