EMAIL THIS PAGE TO A FRIEND
GF90323489 Aldrich

Zirconium

foil, 5m coil, thickness 0.05mm, annealed, 99.2%

Synonym: ZR000220, Zirconium

Purchase

Properties

mfr. no.   Goodfellow, 903-234-89
resistivity   40 μΩ-cm, 20°C
bp   4377 °C (lit.)
mp   1852 °C (lit.)
density   6.5 g/mL at 25 °C (lit.)
SMILES string   [Zr]
InChI   1S/Zr
InChI key   QCWXUUIWCKQGHC-UHFFFAOYSA-N

Description

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?