EMAIL THIS PAGE TO A FRIEND
GF91583264

Rhodium

microfoil, disks, 25mm, thinness 0.25μm, specific density 310μg/cm2, permanent mylar 3.5μm support, 99.99%

Synonym: RH004600, Rhodium

  • CAS Number 7440-16-6

  • Empirical Formula (Hill Notation) Rh

  • Molecular Weight 102.91

  •  MDL number MFCD00011201

  •  PubChem Substance ID 329813787

  •  NACRES NA.23

Purchase

Properties

mfr. no.   Goodfellow, 915-832-64
resistivity   4.33 μΩ-cm, 20°C
bp   3727 °C (lit.)
mp   1966 °C (lit.)
density   12.41 g/cm3 (lit.)
SMILES string   [Rh]
InChI   1S/Rh
InChI key   MHOVAHRLVXNVSD-UHFFFAOYSA-N

Description

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?