• USA Home
  • GF93740384 - Carbon - Vitreous

EMAIL THIS PAGE TO A FRIEND
GF93740384

Carbon - Vitreous

rod, 100mm, diameter 10mm, glassy carbon

Synonym: Carbon - Vitreous, VC007960

Purchase

Properties

form   rod
mfr. no.   Goodfellow, 937-403-84
InChI   1S/C
InChI key   OKTJSMMVPCPJKN-UHFFFAOYSA-N

Description

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Safety & Documentation

Safety Information

WGK Germany 
nwg
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?