EMAIL THIS PAGE TO A FRIEND
GF97050652 Aldrich

Cadmium

foil, 75x100mm, thickness 0.5mm, as rolled, 99.99%

Synonym: CD000261, Cadmium

  • CAS Number 7440-43-9

  • Empirical Formula (Hill Notation) Cd

  • Molecular Weight 112.41

  •  Beilstein Registry Number 8137359

  •  MDL number MFCD00010914

  •  PubChem Substance ID 329814644

Purchase

Properties

InChI Key   BDOSMKKIYDKNTQ-UHFFFAOYSA-N
mfr. no.   Goodfellow, 970-506-52
resistivity   7.27 μΩ-cm, 22°C
bp   765 °C(lit.)
mp   320.9 °C (lit.)
density   8.65 g/mL at 25 °C(lit.)

Description

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?