EMAIL THIS PAGE TO A FRIEND
GF99502836

Dysprosium

wire reel, 25mm, diameter 0.5mm, hard, 99.9%

Synonym: DY005105, Dysprosium

  • CAS Number 7429-91-6

  • Empirical Formula (Hill Notation) Dy

  • Molecular Weight 162.50

  •  MDL number MFCD00010982

  •  PubChem Substance ID 329815031

  •  NACRES NA.23

Purchase

Properties

assay   ≥99.9%
form   wire
mfr. no.   Goodfellow, 995-028-36
resistivity   89 μΩ-cm, 20°C
bp   2567 °C (lit.)
mp   1412 °C (lit.)
density   8.559 g/mL at 25 °C (lit.)
SMILES string   [Dy]
InChI   1S/Dy
InChI key   KBQHZAAAGSGFKK-UHFFFAOYSA-N

Description

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?