EMAIL THIS PAGE TO A FRIEND
I15505 Aldrich

Isobutyraldehyde

98%

Synonym: 2-Methylpropionaldehyde, 2-Methylpropanal

  • CAS Number 78-84-2

  • Linear Formula (CH3)2CHCHO

  • Molecular Weight 72.11

  •  Beilstein Registry Number 605330

  •  EC Number 201-149-6

  •  MDL number MFCD00006980

  •  PubChem Substance ID 329815354

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C1 to C6, Carbonyl Compounds, Chemical Synthesis,
vapor density   2.5 (vs air)
vapor pressure   66 mmHg ( 4.4 °C)
InChI Key   AMIMRNSIRUDHCM-UHFFFAOYSA-N
assay   98%
autoignition temp.   384 °F
expl. lim.   10 %, 25 °F
  2 %, 32 °F
refractive index   n20/D 1.374(lit.)
bp   63 °C(lit.)
mp   −65 °C (lit.)
density   0.79 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2045 3 / PGII
WGK Germany 
1
RTECS 
NQ4025000
Flash Point(F) 
-2.2 °F
Flash Point(C) 
-19 °C

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?