EMAIL THIS PAGE TO A FRIEND
I6709 Aldrich

Iodoacetamide

97%

  • CAS Number 144-48-9

  • Linear Formula ICH2CONH2

  • Molecular Weight 184.96

  •  Beilstein Registry Number 1739080

  •  EC Number 205-630-1

  •  MDL number MFCD00008028

Purchase

Properties

InChI Key   PGLTVOMIXTUURA-UHFFFAOYSA-N
assay   97%
mp   92-95 °C(lit.)

Description

Application

Alkylating reagent for cysteine residues in peptide sequencing. By virtue of reaction with cysteine, it is an irreversible inhibitor of enzymes with cysteine at the active site. It reacts much more slowly with histidine residues, but that activity is responsible for inhibition of ribonuclease.

Enzyme inactivator.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
AC4200000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

I6125 Iodoacetamide, ≥99% (NMR), crystalline

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?