• USA Home
  • JKA10022 - 8arm-PEG10K-SH, tripentaerythritol core

EMAIL THIS PAGE TO A FRIEND
JKA10022 Sigma-Aldrich

8arm-PEG10K-SH, tripentaerythritol core

average Mn 10,000

Synonym: 8arm-PEG-SH, tripentaerythritol core, 8arm-PEG-SH

  • Linear Formula R(O(CH2CH2O)nCH2CH2SH)8

  •  NACRES NA.23

Purchase

Properties

Related Categories 8-arm PEGs, Materials Science, Multi-arm PEGs, PEG Dendrimers and Multi-arm PEGs, Poly(ethylene glycol) and Poly(ethylene oxide),
mol wt   average Mn 10,000
polymer architecture   shape : 8-arm
functionality : homofunctional
reaction suitability  
reactivity: thiol reactive

Description

General description

8arm PEG Thiol. Crosslinks into hydrogels. Selective for thiol groups under mild reaction conditions. 8ARM(TP)-PEG with tripentaerythritol core has a lower polydispersity and higher molecular weight accuracy compared with the generic 8ARM-PEG with a hexaglycerin core.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Applications of Y-Shape Peg Derivatives for Drug Delivery

The immune system protects the body from disease by resisting the invasion of foreign molecules into its cells, for example peptides and proteins are hydrolyzed by proteolytic enzymes; nucleic acids ...
Dr. Hui Zhu
Material Matters, 2016, 11.3
Keywords: Antibiotics, Antitumor agents, Antivirals, Bacterial conjugations, Cancer, Clinical, Degradations, Digestions, Eliminations, Enzyme activity, Genetic, Hormones, Indicators, Ion-exchange chromatography, Metabolism, Oxidations, PEGylations, Pharmaceutical, Phosphorylations, Reductions, Reproduction

Polyethylene Glycol (PEG) Selection Guide

Poly(ethylene glycol) (PEG) is a synthetic, hydrophilic, biocompatible polymer with widespread use in biomedical and other applications. PEGs are synthesized using a ring-opening polymerization of et...
Keywords: Adhesion, Bacterial conjugations, Click chemistry, Molecular biology, PEGylations, Polymerization reactions, Ring-opening polymerization

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?