• USA Home
  • JKA13011 - mPEG6-Propionic acid

EMAIL THIS PAGE TO A FRIEND
JKA13011 Sigma-Aldrich

mPEG6-Propionic acid

  • Linear Formula CH3O(CH2CH2O)6CH2CH2COOH

  •  NACRES NA.23

Purchase

Properties

Related Categories Materials Science, Monofunctional PEGs, Poly(ethylene glycol) and Poly(ethylene oxide), Polymer Science, Polymers,
polymer architecture   shape : linear
functionality : monofunctional
reaction suitability   reagent type: chemical modification reagent
reactivity: amine reactive

Description

General description

Monodisperse PEG. COOH group is stable and can be activated.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Polyethylene Glycol (PEG) Selection Guide

Poly(ethylene glycol) (PEG) is a synthetic, hydrophilic, biocompatible polymer with widespread use in biomedical and other applications. PEGs are synthesized using a ring-opening polymerization of et...
Keywords: Adhesion, Bacterial conjugations, Click chemistry, Molecular biology, PEGylations, Polymerization reactions, Ring-opening polymerization

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?