EMAIL THIS PAGE TO A FRIEND
JKA8017 Sigma-Aldrich

8arm-PEG20K-COOH

hexaglycerol core, average Mn 20,000

Synonym: 8arm-PEG-COOH

  • Linear Formula R(O(CH2CH2O)nCH2COOH)8

  •  NACRES NA.23

Purchase

Properties

Related Categories 8-arm PEGs, Materials Science, Multi-arm PEGs, PEG Dendrimers and Multi-arm PEGs, Poly(ethylene glycol) and Poly(ethylene oxide),
mol wt   average Mn 20,000
polymer architecture   shape : 8-arm
functionality : homofunctional
reaction suitability  
reactivity: amine reactive

Description

General description

8arm PEG Carboxyl. Crosslinks into hydrogels. Carboxyl group binds amino or other acid reactive chemical groups.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Applications of Y-Shape Peg Derivatives for Drug Delivery

The immune system protects the body from disease by resisting the invasion of foreign molecules into its cells, for example peptides and proteins are hydrolyzed by proteolytic enzymes; nucleic acids ...
Dr. Hui Zhu
Material Matters, 2016, 11.3
Keywords: Antibiotics, Antitumor agents, Antivirals, Bacterial conjugations, Cancer, Clinical, Degradations, Digestions, Eliminations, Enzyme activity, Genetic, Hormones, Indicators, Ion-exchange chromatography, Metabolism, Oxidations, PEGylations, Pharmaceutical, Phosphorylations, Reductions, Reproduction

Polyethylene Glycol (PEG) Selection Guide

Poly(ethylene glycol) (PEG) is a synthetic, hydrophilic, biocompatible polymer with widespread use in biomedical and other applications. PEGs are synthesized using a ring-opening polymerization of et...
Keywords: Adhesion, Bacterial conjugations, Click chemistry, Molecular biology, PEGylations, Polymerization reactions, Ring-opening polymerization

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?