EMAIL THIS PAGE TO A FRIEND
L0300001-HH Sigma-Aldrich

ETT Activator

0.25 M

Synonym: 5-(Ethylthio)-1H-tetrazole, 5-(Ethylthio)tetrazole solution

Purchase

Properties

Related Categories ABI 3900, ABI 39X, Activator, Chemical Synthesis, ETT Activator,
InChI Key   GONFBOIJNUKKST-UHFFFAOYSA-N
type   for DNA synthesis
product line   Proligo Reagents
assay   32-35 g/L (titration)
packaging   pkg of 4 × 2 L L0302001-04
  pkg of 4 × 2.5 L L0302501-04
impurities   ≤30 ppm water content (Karl Fischer)

Description

Preparation Note

Acetonitrile / 5-(Ethylthio)-1H-tetrazole 100 / 3 v/w

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993BE 3 / PGII
WGK Germany 
3
Flash Point(F) 
35.6 °F
Flash Point(C) 
2 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?