• USA Home
  • L511765 - [Ir{dF(CF3)ppy}2(dtbpy)]PF6

EMAIL THIS PAGE TO A FRIEND
L511765 Aldrich

[Ir{dF(CF3)ppy}2(dtbpy)]PF6

AldrichCPR

  • Empirical Formula (Hill Notation) C42H34F16IrN4P

  • Molecular Weight 1121.91

Purchase

Description

Application

Learn More at the Professor and Product Portal of Professor Teshik Yoon and Professor Corey Stephenson.

Versatile visible-light photocatalyst used by Yoon for periselective [2+2] cycloadditions and Stephenson for Atom Transfer Radical Addition of alkyl halides to alkenes and alkynes.

Visible Light Photocatalysis of [2+2] Styrene Cycloadditions by Energy Transfer

Visible Light-Mediated Atom Transfer Radical Addition via Oxidative and Reductive Quenching of Photocatalysts

For other Visible-Light Photocatalysts, refer to: www.aldrich.com/photocatalysis

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Stephenson Group – Professor Product Portal

Professor Corey Stephenson Research in the Stephenson lab focuses upon the development of new chemical methods which enable the activation of chemical bonds under mild reaction conditions. Specifical...
Keywords: Catalysis, Infrared spectroscopy, Materials Science, Natural product synthesis

Yoon Group – Professor Product Portal

From our library of Articles, Sigma-Aldrich presents Yoon Group – Professor Product Portal
Keywords: Cycloadditions, Ligands, Oxidations

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?


Screeing simplified