• USA Home
  • L511889 - 2-Bromo-6-iodobenzoic acid

EMAIL THIS PAGE TO A FRIEND
L511889 Sigma-Aldrich

2-Bromo-6-iodobenzoic acid

AldrichCPR

Purchase

Properties

Related Categories Building Blocks, C-H Activation, Catalysis and Inorganic Chemistry, Chemical Synthesis
Quality Level   100
reaction suitability   reagent type: ligand
reaction type: C-H Activation
SMILES string   OC(C1=C(Br)C=CC=C1I)=O
InChI   1S/C7H4BrIO2/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3H,(H,10,11)
InChI key   PTQGEYMPMWQTDM-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?