EMAIL THIS PAGE TO A FRIEND
L512125 Sigma-Aldrich

DPAVBi

AldrichCPR

Synonym: 4,4′ -Bis[4-(di-p-tolylamino)styryl]biphenyl

Purchase

Properties

Related Categories Light Emitters and Dopants, Materials Science, OLED and PLED Materials, Organic and Printed Electronics
InChI Key   OSQXTXTYKAEHQV-WXUKJITCSA-N

Description

General description

DPAVBi is a blue fluorescent dopant material. It exhibits a maximum electroluminescent efficiency of 8.6 cd/A 7.4 cd/A at 1000 cd/m2 and 10.4 lm/W at 2.6 V 6.4 lm/W at 1000 cd/m2.

Application

It may be used as a dopant for fluorescent host material, 6-anthracene-9-yl-2,3-di-p-tolyl benzo[b]thiophene (ATB) for the fabrication of organic light emitting diodes (OLEDs.) It may also be used as a dopant for white organic light emitting diodes.

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?