EMAIL THIS PAGE TO A FRIEND
S6525 Aldrich

Sulfanilamide

99%

Synonym: p-Aminobenzenesulfonamide

  • CAS Number 63-74-1

  • Linear Formula H2NC6H4SO2NH2

  • Molecular Weight 172.20

  •  Beilstein Registry Number 511852

  •  EC Number 200-563-4

  •  MDL number MFCD00007939

Purchase

Properties

InChI Key   FDDDEECHVMSUSB-UHFFFAOYSA-N
assay   99%
mp   164-166 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
WO8400000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

S9251 Sulfanilamide, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?