EMAIL THIS PAGE TO A FRIEND
SFL00001

PhenoFluor

AldrichCPR

Synonym: 1,3-Bis(2,6-diisopropylphenyl)-2,2-difluoro-2,3-dihydro-1H-imidazole

  • CAS Number 1314657-40-3

  • Empirical Formula (Hill Notation) C27H36F2N2

  • Molecular Weight 426.58

  •  MDL number MFCD22377831

Purchase

Properties

storage temp.   −20°C
SMILES string   FC1(F)N(C2=C(C(C)C)C=CC=C2C(C)C)C=CN1C3=C(C(C)C)C=CC=C3C(C)C

Description

Application

PhenoFluor reagent is utilized in deoxyfluorination of phenols, without activation of the substrate, to provide ipso substitution to the corresponding aryl fluoride.

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Legal Information

PhenoFluor is a trademark of SciFluor Life Sciences, LLC

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?