• USA Home
  • SYX00051 - 2-(Trifluoromethyl)-1,3-bis(dimethylamino)trimethinium hexafluorophosphate

EMAIL THIS PAGE TO A FRIEND
SYX00051 Sigma-Aldrich

2-(Trifluoromethyl)-1,3-bis(dimethylamino)trimethinium hexafluorophosphate

AldrichCPR

Purchase

Properties

Related Categories Amines, Building Blocks, C8, Chemical Synthesis, Fluorinated Building Blocks,
SMILES string   F[P-](F)(F)(F)(F)F.CN(C)\C=C(\C=[N+](\C)C)C(F)(F)F
InChI   1S/C8H14F3N2.F6P/c1-12(2)5-7(6-13(3)4)8(9,10)11;1-7(2,3,4,5)6/h5-6H,1-4H3;/q+1;-1
InChI key   GMWIFDFSXGELQO-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Legal Information

Product of Synthonix

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?