EMAIL THIS PAGE TO A FRIEND
T57606 Aldrich

Tridecanoic acid

98%

  • CAS Number 638-53-9

  • Linear Formula CH3(CH2)11CO2H

  • Molecular Weight 214.34

  •  Beilstein Registry Number 508317

  •  EC Number 211-341-1

  •  MDL number MFCD00002741

  •  PubChem Substance ID 329826830

Purchase

Properties

InChI Key   SZHOJFHSIKHZHA-UHFFFAOYSA-N
assay   98%
bp   236 °C/100 mmHg(lit.)
mp   41-42 °C(lit.)
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
YD3850000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

T0502 Tridecanoic acid, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?