EMAIL THIS PAGE TO A FRIEND
W215317 Sigma-Aldrich

Bergamot oil

Ivory Coast origin, (Non-sensitizing)

Purchase

Properties

biological source   synthetic
reg. compliance   FDA 21 CFR 182.20
origin   Ivory Coast origin
refractive index   n20/D 1.4665 (lit.)
  n20/D 1.4675 (lit.)
bp   159 °C (lit.)
density   0.877 g/mL at 25 °C (lit.)
Documentation   see Safety & Documentation for available documents
Organoleptic   fruity; sweet
food allergen   no known allergens

Description

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
2
RTECS 
DS0500000
Flash Point(F) 
136.4 °F
Flash Point(C) 
58 °C
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?