• USA Home
  • W250309 - Garlic oil, Chinese

EMAIL THIS PAGE TO A FRIEND
W250309 Sigma-Aldrich

Garlic oil, Chinese

artificial, FCC, FG

Synonym: Garlic oil

Purchase

Properties

Related Categories Alphabetical Listings, Certified Food Grade Products, E - H, E-G, E-H,
grade   FG
  Halal
  Kosher
reg. compliance   FCC
  FDA 21 CFR (117)
  EU Regulation 1334/2008 & 178/2002
origin   China origin
refractive index   n20/D 1.575
density   1.083 g/mL at 25 °C
Documentation   see Safety & Documentation for available documents
Organoleptic   garlic; alliaceous
food allergen   no known allergens

Description

Packaging

1 kg in glass bottle

5, 10 kg in steel drum

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
RTECS 
LX3154800
Flash Point(F) 
116.6 °F
Flash Point(C) 
47 °C
Protocols & Articles

Articles

Securing a Traceable Supply Chain for Food Grade Flavor Ingredients

Introduction Defining "Food Grade" Understanding "Food Grade" Versus "Industrial Grade" Top Questions to Ask a Flavor & Fragrances Supplier The Sigma-Aldrich Approach
Dr. Luke Grocholl
Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Food & Beverage, Food Safety, Solvents

Understanding the Complexities of Kosher Ingredients

Introduction What is Kosher Product Considerations for Kosher Certification Kosher's Added Benefit to the Food Industry Sigma-Aldrich's Kosher Experience and Offering
Dr. Luke Grocholl1, Rabbi Gershon Segal

1Dr. Luke Grocholl, Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food & Beverage, Safety industry

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food
06732 Timestrip® Seafood

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?