EMAIL THIS PAGE TO A FRIEND
W274607 Aldrich

Dimethyl sulfide

≥99%, FCC

Synonym: DMS, Methyl sulfide

  • CAS Number 75-18-3

  • Linear Formula (CH3)2S

  • Molecular Weight 62.13

  •  FEMA Number 2746

  •  Beilstein Registry Number 1696847

  •  EC Number 200-846-2

  •  Council of Europe no. 483

  •  MDL number MFCD00008562

  •  PubChem Substance ID 24901304

  •  Flavis number 12.006

Purchase

Properties

biological source   synthetic
reg. compliance   FCC
vapor density   2.1 (vs air)
vapor pressure   26.24 psi ( 55 °C)
  7.79 psi ( 20 °C)
InChI Key   QMMFVYPAHWMCMS-UHFFFAOYSA-N
assay   ≥99%
autoignition temp.   402 °F
expl. lim.   19.7 %
refractive index   n20/D 1.435(lit.)
bp   38 °C(lit.)
mp   −98 °C (lit.)
density   0.846 g/mL at 25 °C(lit.)
Gene Information   rat ... Maoa(29253), Maob(25750)

Description

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

RIDADR 
UN 1164 3/PG 2
WGK Germany 
1
RTECS 
PV5075000
Flash Point(F) 
-33 °F
Flash Point(C) 
-36 °C
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?