EMAIL THIS PAGE TO A FRIEND
W282529 Aldrich

Orange oil

Florida origin

Synonym: Citrus aurantium, Citrus sinensis

Purchase

Properties

Related Categories Alphabetical Listings, Essential Oils, Flavors and Fragrances, Kosher Certified Products, O-P,
biological source   Citrus sinensis (L.) Osbeck
grade   Kosher
optical activity   [α]20/D +116°, neat
origin   Florida origin
refractive index   n20/D 1.473
bp   176 °C
density   0.845 g/mL at 25 °C
Documentation   see Safety & Documentation for available documents
Organoleptic   orange
food allergen   no known allergens

Description

Preparation Note

Extraction method: cold pressed

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
2
RTECS 
RI8600000
Flash Point(F) 
129.2 °F
Flash Point(C) 
54 °C
Protocols & Articles

Articles

Understanding the Complexities of Kosher Ingredients

Introduction What is Kosher Product Considerations for Kosher Certification Kosher's Added Benefit to the Food Industry Sigma-Aldrich's Kosher Experience and Offering
Dr. Luke Grocholl1, Rabbi Gershon Segal

1Dr. Luke Grocholl, Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food & Beverage, Safety industry

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food
06732 Timestrip® Seafood

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?