EMAIL THIS PAGE TO A FRIEND
W303305 Sigma-Aldrich

Spike lavender oil

Synonym: Lavender spike oil

Purchase

Properties

Related Categories Alphabetical Listings, Essential Oils, Flavors and Fragrances, Kosher Certified Products, O-Z,
biological source   Lavandula spp.
grade   Kosher
optical activity   [α]20/D −2.5°, neat
origin   Spain origin
refractive index   n20/D 1.47 (lit.)
bp   183 °C(lit.)
density   0.893 g/mL at 25 °C (lit.)
Documentation   see Safety & Documentation for available documents
Organoleptic   floral; vanilla

Description

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
134.6 °F
Flash Point(C) 
57 °C

New Product Offerings
Protocols & Articles

Articles

Understanding the Complexities of Kosher Ingredients

Introduction What is Kosher Product Considerations for Kosher Certification Kosher's Added Benefit to the Food Industry Sigma-Aldrich's Kosher Experience and Offering
Dr. Luke Grocholl1, Rabbi Gershon Segal

1Dr. Luke Grocholl, Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food & Beverage, Safety industry

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?