EMAIL THIS PAGE TO A FRIEND
W322903 Aldrich

Isopropyl tiglate

≥98%

  • CAS Number 1733-25-1

  • Linear Formula CH3CH=C(CH3)CO2CH(CH3)2

  • Molecular Weight 142.20

  •  FEMA Number 3229

  •  EC Number 217-067-9

  •  Council of Europe no. 10733

  •  MDL number MFCD00016631

  •  PubChem Substance ID 24901569

  •  Flavis number 9.513

Purchase

Properties

Related Categories Alphabetical Listings, Flavors and Fragrances, Halal Certified Products, I - N, I-L,
biological source   synthetic
grade   Halal
  Kosher
InChI Key   VUPBIVVRPJDWNW-FNORWQNLSA-N
assay   ≥98%
refractive index   n20/D 1.431(lit.)
density   0.896 g/mL at 25 °C(lit.)
Documentation   see Safety & Documentation for available documents
Organoleptic   minty

Description

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3272 3 / PGIII
WGK Germany 
2
RTECS 
EM9254500
Flash Point(F) 
123.8 °F
Flash Point(C) 
51 °C
Protocols & Articles

Articles

Understanding the Complexities of Kosher Ingredients

Introduction What is Kosher Product Considerations for Kosher Certification Kosher's Added Benefit to the Food Industry Sigma-Aldrich's Kosher Experience and Offering
Dr. Luke Grocholl1, Rabbi Gershon Segal

1Dr. Luke Grocholl, Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food & Beverage, Safety industry

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food
06732 Timestrip® Seafood

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?