EMAIL THIS PAGE TO A FRIEND
W338001 Sigma-Aldrich

Linoleic acid

≥95%

Synonym: 9-cis,12-cis-Linoleic acid, cis-9,cis-12-Octadecadienoic acid, Telfairic acid

  • CAS Number 60-33-3

  • Linear Formula CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H

  • Molecular Weight 280.45

  •  Beilstein/REAXYS Number 1727101

  •  EC Number 200-470-9

  •  Council of Europe no. 694

  •  MDL number MFCD00064241

  •  PubChem Substance ID 24901678

Purchase

Properties

biological source   peanut oil
  synthetic
assay   ≥95%
form   liquid
refractive index   n20/D 1.466 (lit.)
bp   229-230 °C/16 mmHg (lit.)
mp   −5 °C (lit.)
density   0.902 g/mL at 25 °C (lit.)
Documentation   see Safety & Documentation for available documents
Organoleptic   faint; fatty
food allergen   peanuts
SMILES string   OC(CCCCCCC/C=C\C/C=C\CCCCC)=O
InChI   1S/C18H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h6-7,9-10H,2-5,8,11-17H2,1H3,(H,19,20)/b7-6-,10-9-
InChI key   OYHQOLUKZRVURQ-HZJYTTRNSA-N
Gene Information   human ... SOAT1(6646), SOAT2(8435)

Description

General description

Linoleic acid is a polyunsaturated essential fatty acid that is mainly found in vegetable oils such as sunflower, corn, soy, and groundnut oil.

Packaging

25, 100 g in glass bottle

Biochem/physiol Actions

Linoleic acid increases cell proliferation and gene expression of PPARα and its target genes such as acyl-CoA oxidase in primary duck hepatocytes .

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
RF9990000
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles

Articles

Omega-3 Fatty Acids and Heart Disease

The potential for the prevention and treatment of cardiovascular disease through increased dietary intake of omega-3 (w-3) fish oils is not a recent scientific discovery. A historically important stu...
BioFiles 2007, 2.2, 14.
Keywords: Anti-inflammatory agents, Biochemistry, Cardiovascular, Clinical, Degradations, Esterifications, Immunology, Inflammation, Lipid Metabolism, Metabolism, Metabolites, Methods, Nutrition, Oxidations, Transcription

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?