• USA Home
  • W352705 - 3-Acetyl-2,5-dimethylthiophene

EMAIL THIS PAGE TO A FRIEND
W352705 Aldrich

3-Acetyl-2,5-dimethylthiophene

≥99%

  • CAS Number 2530-10-1

  • Empirical Formula (Hill Notation) C8H10OS

  • Molecular Weight 154.23

  •  EC Number 219-779-5

  •  Council of Europe no. 11603

  •  MDL number MFCD00009763

  •  PubChem Substance ID 24901762

  •  Flavis number 15.024

Purchase

Properties

Related Categories A - D, A-B, A-D, Alphabetical Listings, Flavors and Fragrances,
biological source   synthetic
grade   Halal
  Kosher
InChI Key   PUSJAEJRDNPYKM-UHFFFAOYSA-N
assay   ≥99%
refractive index   n20/D 1.544(lit.)
bp   105-108 °C/15 mmHg(lit.)
density   1.086 g/mL at 25 °C(lit.)
Documentation   see Safety & Documentation for available documents
Organoleptic   meaty
food allergen   no known allergens

Description

Application

Effective June 2013, FEMA and the European Union have removed 3-Acetyl-2,5-dimethylthiophene from the FEMA GRAS and Union List of flavoring substances as suitable for use in food as a flavoring ingredient. Pending additional review of safety data, a final determination will be made by those respective organizations regarding the safe use of this substance.

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
UN 3334
WGK Germany 
3
Flash Point(F) 
210.2 °F
Flash Point(C) 
99 °C
Protocols & Articles

Articles

Understanding the Complexities of Kosher Ingredients

Introduction What is Kosher Product Considerations for Kosher Certification Kosher's Added Benefit to the Food Industry Sigma-Aldrich's Kosher Experience and Offering
Dr. Luke Grocholl1, Rabbi Gershon Segal

1Dr. Luke Grocholl, Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food & Beverage, Safety industry

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?


New Product Offerings