EMAIL THIS PAGE TO A FRIEND
W502944 Aldrich

Cinnamyl nitrile

Synonym: 3-Phenylacrylonitrile, Cinnamonitrile

  • CAS Number 1885-38-7

  • Linear Formula C6H5CH=CHCN

  • Molecular Weight 129.16

  •  Beilstein Registry Number 1209546

  •  EC Number 217-552-5

  •  MDL number MFCD00001930

Purchase

Properties

InChI Key   ZWKNLRXFUTWSOY-QPJJXVBHSA-N
refractive index   n20/D 1.601(lit.)
  n20/D 1.602
bp   254-255 °C(lit.)
mp   18-20 °C(lit.)
  18-20 °C
density   1.028 g/mL at 25 °C(lit.)
  1.028 g/mL
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
2
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

New F&F Product Supplement
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

C81004 Cinnamonitrile, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?