EMAIL THIS PAGE TO A FRIEND
W506206 Sigma-Aldrich

2,4-Xylenol

≥98%

Synonym: 2,4-Dimethylphenol, 4-Hydroxy-m-xylene, asym.-m-Xylenol

  • CAS Number 105-67-9

  • Linear Formula (CH3)2C6H3OH

  • Molecular Weight 122.16

  •  Beilstein/REAXYS Number 636244

  •  EC Number 203-321-6

  •  MDL number MFCD00002233

  •  PubChem Substance ID 24902027

  •  Flavis number 4.066

  •  NACRES NA.21

Purchase

Properties

Related Categories Alphabetical Listings, Flavors and Fragrances, Q-Z More...
Quality Level   400
biological source   synthetic
grade   Halal
Agency/Method   follows IFRA guidelines
reg. compliance   EU Regulation 1223/2009
vapor pressure   0.1 mmHg ( 25 °C)
assay   ≥98%
refractive index   n20/D 1.538 (lit.)
bp   211-212 °C (lit.)
mp   22-23 °C (lit.)
density   1.011 g/mL at 25 °C (lit.)
Documentation   see Safety & Documentation for available documents
Organoleptic   burnt; roasted
food allergen   no known allergens
fragrance allergen   no known allergens
storage temp.   room temp
SMILES string   Cc1ccc(O)c(C)c1
InChI   1S/C8H10O/c1-6-3-4-8(9)7(2)5-6/h3-5,9H,1-2H3
InChI key   KUFFULVDNCHOFZ-UHFFFAOYSA-N

Description

Packaging

1 kg in glass bottle

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2261 6.1 / PGII
WGK Germany 
WGK 3
RTECS 
ZE5600000
Flash Point(F) 
201.2 °F - closed cup
Flash Point(C) 
94.0 °C - closed cup
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?