EMAIL THIS PAGE TO A FRIEND
W524409 Sigma-Aldrich

Trivertal

≥98%

  • CAS Number 27939-60-2

  • Empirical Formula (Hill Notation) C9H14O

  • Molecular Weight 138.21

  •  FEMA Number 4505

  •  EC Number 248-742-6

  •  PubChem Substance ID 24902100

  •  NACRES NA.21

Purchase

Properties

Related Categories Alphabetical Listings, Flavors and Fragrances, Q-Z More...
Quality Level   400
biological source   synthetic
assay   ≥98%
refractive index   n20/D 1.473 (lit.)
bp   196 °C (lit.)
density   0.936 g/mL at 25 °C (lit.)
Documentation   see Safety & Documentation for available documents
Organoleptic   green; herbaceous
food allergen   no known allergens
SMILES string   CC1CC(CC(C)=C1)C=O
InChI   1S/C9H14O/c1-8-5-3-4-6-9(8,2)7-10/h3,5,7-8H,4,6H2,1-2H3
InChI key   CYGGTUUCLIFNIU-UHFFFAOYSA-N

Description

Packaging

1 kg in glass bottle

10 kg in composite drum

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

WGK Germany 
WGK 2
Flash Point(F) 
303.8 °F - closed cup
Flash Point(C) 
151 °C - closed cup
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?