• USA Home
  • WHA10407992 - Whatman® ME membranes for microbiological control
EMAIL THIS PAGE TO A FRIEND
WHA10407992
Whatman® ME membranes for microbiological control
ME 25/91, diam. 50 mm, pore size 0.45 μm, pack of 100
Purchase
Description
General description
These membranes are used to quantitatively evaluate microorganism counts. The liquid is filtered through a membrane and microorganisms collect on the membrane surface. The filter is incubated on a nutrient medium and the individual colonies can then be evaluated. The method is also suitable for large sample volumes and low microorganism counts.
• Mixed cellulose ester membrane
• Economical
• Specifically for aqueous solutions
• Hydrophilic
• For use up to 125ºC
Legal Information
Whatman is a registered trademark of Cytiva
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?