• USA Home
  • WHA68692502 - Whatman® GD/X syringe filters

EMAIL THIS PAGE TO A FRIEND
WHA68692502

Whatman® GD/X syringe filters

Nylon membrane, pore size 0.2 μm, diam. 25 mm, non-sterile, pkg of 150 ea

Synonym: Whatman filter, Z745235, syringe filter, syringe filters

  •  NACRES NB.24

Purchase

Properties

Related Categories GD/X syringe filters, HPLC Sample Preparation, Labware, Manufacturer Browser, Whatman,
material   Nylon membrane
  polypropylene housing
sterility   non-sterile
feature   glass fiber prefilter
packaging   pkg of 150 ea
mfr. no.   Whatman Article No., 28420487 (US reference)
  Whatman, 6869-2502
diam.   25 mm
volume   ≤25 mL
pore size   0.2 μm
fitting   female Luer inlet (lock)
  male Luer outlet

Description

General description

The GD/X filter contains a unique prefiltration stack of 100% borosilicate glass. The prefilter will retain fine particles down to 0.7 μm, allowing difficult sample filtration with less hand force.

The volume of the sample filtered is 3-7 times greater than an unprotected membrane. PP housing. Syringe filters can be autoclaved.

Membranes:

Cellulose Acetate:
for biological solutions, high-loading capacity, and low-protein-binding
Nylon: for aqueous and aqueous organics
Polyethersulfone: for serum, plasma, and tissue culture solutions
Polypropylene: high flow for aqueous and organics
Polysulfone: low-protein-adsorption for serums and plasma
PTFE: chemically inert, will stop aqueous aerosols in gas streams
PVDF: the most suitable membrane for HPLC
Glass microfiber: borosilicate with high-load capacity

Suitability

Suitable for filtering environmental and food analysis samples. Suitable for particulate laden sample preparation.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Whatman® GD/X Syringe Filters [VIDEO]

These innovative syringe filters are specially designed for the handling of high-particulate solutions. The specialized membrane consists of 4-layers of high-quality Whatman filtration media, which a...
Keywords: Filtration

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?