• USA Home
  • Z118168 - Straight septum-inlet adapter
EMAIL THIS PAGE TO A FRIEND
Z118168 Sigma-Aldrich
Straight septum-inlet adapter
inlet I.D. 6 mm, stainless steel
  •  NACRES NB.43
Purchase
Description
General description
1/4 in. NPTM × inlet 6 mm I.D. adapters require septa Z565687 or Z565695.
1/4 in. NPTM × inlet 13.9 mm I.D. adapters require septa Z124591 or Z167282
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Articles
Pressure-Temperature Calculator for Solvents
*Calculations provide estimated results based on Antoine's Coefficients and their associated temperature range values. Antoine's Coefficients were obtained from the National Institute of Standards an...
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?