• USA Home
  • Z118192 - Straight septum-inlet adapter
EMAIL THIS PAGE TO A FRIEND
Z118192 Sigma-Aldrich
Straight septum-inlet adapter
inlet I.D. 13.9 mm, brass
  •  NACRES NB.43
Purchase
Description
General description
1/4 in. NPTM × inlet 6 mm I.D. adapters require septa Z565687 or Z565695.
1/4 in. NPTM × inlet 13.9 mm I.D. adapters require septa Z124591 or Z167282
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Articles
Pressure-Temperature Calculator for Solvents
*Calculations provide estimated results based on Antoine's Coefficients and their associated temperature range values. Antoine's Coefficients were obtained from the National Institute of Standards an...
Related Content
Properties of Solvents Table
The table below presents the physical characteristics of common solvents. Easily reference or compare solvent properties like boiling point, density, and refractive index all from one list.
Keywords: Gas chromatography, Green chemistry, High performance liquid chromatography, Melting, Solvents
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?