• USA Home
  • Z124907 - Contemporary Heterocyclic Chemistry. Syntheses, Reactions, and Applications
EMAIL THIS PAGE TO A FRIEND
Z124907 Aldrich
Contemporary Heterocyclic Chemistry. Syntheses, Reactions, and Applications
  •  ISBN-10 0471062790
  •  ISBN-13 978-047106279-0
Purchase
Properties
publication info   G.R. Newkome, John Wiley & Sons, Ltd, 1982, hard cover
mfr. no.   Wiley
Description
General description
A comprehensive approach to heterocyclic chemistry, based on the concept of PI [ -excessive and PI [ -deficient ring systems. Introduces the categorical similarities and differences among the varied heterocyclic systems. Explains the logical consistency of the numerous systhesis and relative reactivities of heterocyclic compounds, as demonstrated not only by their chemical reactions, but by their spectroscopic properties as well. Also provides highly useful tabulation of nuclear magnetic resonance data for proton, carbon and nitrogen, mass spectral fragmentation, and the results of unified molecular orbital calculations.
Table of Contents
PARTIAL TABLE OF CONTENTS:
Nomenclature.
Electronic Division of Aromatic Heterocyclic Compounds.
Synthetic Routes to Selected Monocyclic Five-Membered Pi-Excessive Heteroaromatic Compounds.
Synthetic Routes and Reactions of Some Common Benzo-fused Five Membered Pi-Excessive Heteroaromatic Compounds.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?