• USA Home
  • Z225657 - Aldrich® Benchrack portable lattice system
EMAIL THIS PAGE TO A FRIEND
Z225657 Sigma-Aldrich
Aldrich® Benchrack portable lattice system
mini-size, 4 positions, 8 rods
  •  NACRES NB.35
Purchase
Description
General description
Includes base and 1/2 in. diam. lattice rod sections.
Portable lattice systems fit easily on a bench or in a fume hood and may be disassembled for storage or transport. Solid aluminum lattice rod sections, 11 3/4 in. L, screw into each other for extra height. Nonslip rubber feet on base. Entire system assembles without tools.
For more permanent setups, machined flats on lattice rods may be tightened with a wrench (included). Available in two standard sizes. Also available in a mini-size for use with small and microscale glassware and in medium and large size Aldrich AtmosBags.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Benchrack is a trademark of Sigma-Aldrich Co. LLC
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
recommended
Product #
Description
Add to Cart
Z106089 Aldrich® AtmosBag, two-hand, non-sterile, size L, closure type, Tape-seal
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?