• USA Home
  • Z225665 - Aldrich® Benchrack portable lattice rod only
EMAIL THIS PAGE TO A FRIEND
Z225665 Sigma-Aldrich
Aldrich® Benchrack portable lattice rod only
rod diam. × L 5/8  in. × 11.75 in., Standard
  •  NACRES NB.35
Purchase
Description
General description
Rod sections with threaded ends screw together, building extra height to Benchrack systems.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Benchrack is a trademark of Sigma-Aldrich Co. LLC
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?