• USA Home
  • Z252999 - Enviro-Safe® thermometer
EMAIL THIS PAGE TO A FRIEND
Z252999
Enviro-Safe® thermometer
Green Alternative
temperature -20-110 °C, immersion level, total
  •  NACRES NB.79
Purchase
Properties
Related Categories 12 Principles Aligned Products, Chemical Synthesis, Enviro-Safe thermometer, Greener Alternative Products, HB Instruments,
feature   immersion level total
mfr. no.   HB Instruments 20500
greener alternative product characteristics   Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.
temperature measuring range   -20-110 °C
L   305 mm
Description
General description
Sigma Life Science is committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is designed as a safer chemicals without mercury. Find details here.
Enviro-Safe thermometers are serialized, calibrated with NIST (NBS) traceable standards, and include a Statement of Accuracy.
White glass with easy-to-read, non-hazardous, biodegradable, green liquid fill. 6.5 mm O.D. guaranteed accuracy: ±1 scale division below 105°C (221°F), ±1.5 scale divisions above 105°C (221°F), and ±2 scale divisions above 200°C (392 °F).
Legal Information
Enviro-Safe is a registered trademark of H-B Instrument Co.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
92451 Timestrip Plus 25 °C
08168 Timestrip Plus 8 °C
06693 Timestrip Plus -20 °C
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?