• USA Home
  • Z258725 - Ace vacuum-sublimation apparatus
EMAIL THIS PAGE TO A FRIEND
Z258725 Sigma-Aldrich
Ace vacuum-sublimation apparatus
Repl. CAPFE O-ring for Z258709
Purchase
Description
General description
Features a greaseless O-ring sealed flange with a quick-release clamp. Top flange is ground flat, bottom mating flange has a groove for a CAPFE (fluororesin encapsulated silicone) O-ring. Complete apparatus includes an outer body, inner top, CAPFE O-ring, and quick-release clamp.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?