• USA Home
  • Z263885 - Carbon-13 NMR Spectroscopy Of Biological Systems
EMAIL THIS PAGE TO A FRIEND
Z263885
Carbon-13 NMR Spectroscopy Of Biological Systems
  •  ISBN-10 0120843706
  •  ISBN-13 978-012084370-1
Purchase
Properties
publication info   N. Beckmann, ed., Academic Press, 1995, 334 pp., hard cover
mfr. no.   Elsevier
Description
General description
This book is intended to provide an in-depth understanding of 13C NMR as a tool in biological research. 13C NMR has provided unique information concerning complex biological systems, from proteins and nucleic acids to animals and humans. It is not a push-button technique, and its sound application in biological research requires a strong interaction between individuals with different backgrounds. The contents have been chosen to be of interest to biochemists, biophysicists, chemists, medical researchers, molecular biologists, and physicists. The book emphasizes both the NMR methodology itself and the motivation for the use of 13C NMR to address certain biological problems.
Table of Contents
Introduction.
Methodology and Applications of Heteronuclear and Multidimensional 13C NMR to the Elucidation of Molecular Structure and Dynamics in the Liquid State.
Structural Determination of Biological Solids by Magic-Angle Spinning 13C NMR.
13C NMR Studies of the Interactions of Fatty Acids with Phospholipid Bilayers, Plasma Lipoproteins, and Proteins.
Application 13C NMR Spectroscopy to Metabolic Studies on Animals.
13C Magnetic Resonance Spectroscopy as a Noninvasive Tool for Metabolic Studies on Humans.
Subject Index.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?