• USA Home
  • Z268488 - Gelman Acro 50 vent filter
EMAIL THIS PAGE TO A FRIEND
Z268488 Sigma-Aldrich
Gelman Acro 50 vent filter
PTFE membrane, diam. 50 mm, pore size 0.2 μm
Purchase
Properties
material   PTFE membrane
  polypropylene housing
sterility   non-sterile
packaging   pkg of 18
mfr. no.   Pall 4251
diam.   50 mm
pore size   0.2 μm
Description
General description
Reusable 50 mm filters with superior performance for critical applications.
• Reusable, autoclavable. Ideal for use with bioreactors and fermenters
• PTFE membrane and polypropylene housing have broad chemical resistance
• Large surface area provides greater throughput
Suitability
Suitable for venting bioreactors, fermentation tanks, and carboys.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?