• USA Home
  • Z283908 - Spinbar® magnetic stir bar

EMAIL THIS PAGE TO A FRIEND
Z283908

Spinbar® magnetic stir bar

round PTFE-coated, size 30 mm × 9 mm, white

  •  NACRES NB.72

Purchase

Properties

Related Categories 21mm to 40mm length, Bel-Art, Bel-Art Magnetic Stirring Bars, Bel-Art Spinbar Magnetic Stirring Bars, Cylindrical Stir Bars,
material   polygon
  round PTFE-coated
description   Tapered design acts as pivot point
mfr. no.   Bel-Art, F37113-0309
  Bel-Art
L   30 mm
diam.   9 mm
size   30 mm × 9 mm
color   white

Description

General description

Leakproof, seamless, polished, and contains a permanent Alnico V magnet. Egg-shape bar is designed especially for use in round-bottom vessels.

Legal Information

Spinbar is a registered trademark of SP Scienceware, a division of SP

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?