• USA Home
  • Z307653 - Primeguard® standard coveralls
EMAIL THIS PAGE TO A FRIEND
Z307653 Aldrich
Primeguard® standard coveralls
size XXL
Purchase
Properties
size   XXL
availability   not available in USA and Canada
Description
Features and Benefits
With integral hood, full length zipper and elasticated wrist and ankle holes. Soft, comfortable fabric with textile feel and appearance. Provides CE Type 5 and Type 6 (particle and limited splash) protection.
Legal Information
Primeguard is a registered trademark of Shiloh Healthcare Ltd.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?