• USA Home
  • Z311847 - Mettler Toledo PB precision balance, 10 mg readability range
EMAIL THIS PAGE TO A FRIEND
Z311847 Aldrich
Mettler Toledo PB precision balance, 10 mg readability range
Model PB1502-S, AC/DC input 230 V AC
Purchase
Properties
feature   CE compliant
mfr. no.   Model PB1502-S
availability   available only in EU
measuring range   1510 g weighing capacity, precision: 10 mg, resolution: 10 mg
weighing pan diam.   180 mm
Description
Features and Benefits
• Calibration: Using external weight
  • DeltaRange (DR) is a Mettler-Toledo feature that ensures the most accurate readability mode is used at all times. DeltaRange models have a fine range of 10 times better readability than the normal range. The fine range starts every time the balance is tared (for example, after placing a container on the balance). The balance automatically reverts to normal readability after the fine range is exceeded. DeltaRange can be recalled again at any time. See balances with DR as a suffix in the model number.
  • Calibration weights, RS232 interface, BalanceLink software, printers and accessories available separately.
  • Safety & Documentation
    Safety Information
    Safety Information for this product is unavailable at this time.
    Documents
    Certificate of Analysis
    Protocols & Articles
    Related Products
    Technical Service:
    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
    Bulk Ordering & Pricing:
    Need larger quantities for your development, manufacturing or research applications?