EMAIL THIS PAGE TO A FRIEND
Z313882 Aldrich
Retsch test sieves
4 mm, D × H 200 mm × 50 mm
Purchase
Properties
material   stainless steel (with wire gauze)
availability   not available in USA and Canada
D × H   200 mm × 50 mm
particle size   4 mm
Description
General description
For mechanical or manual, wet or dry sieving. Quintuple tested and provided with Quality Certificate DIN ISO 3310/1.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?