• USA Home
  • Z417947 - Aldrich® System 45 Kjeldahl flask
EMAIL THIS PAGE TO A FRIEND
Z417947 Sigma-Aldrich
Aldrich® System 45 Kjeldahl flask
thread size, 45 mm, capacity 1,000 mL
Purchase
Properties
feature   thread size 45 mm
capacity   1,000 mL
Description
General description
Single-neck with 32 or 45 mm threaded top. This design allows access to sample with a spatula or policeman. Flasks may be easily stored with screw-cap closures in place. Flask fits standard heating mantles. Cap sold separately.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
System 45 is a trademark of John Babashak
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?