• USA Home
  • Z422398 - Sax’s Dangerous Properties of Industrial Materials, 10th Edition, 3 Vol. Set

EMAIL THIS PAGE TO A FRIEND
Z422398 Aldrich

Sax’s Dangerous Properties of Industrial Materials, 10th Edition, 3 Vol. Set

  •  ISBN-10 0471354074

  •  ISBN-13 978-047135407-9

Purchase

Properties

publication info   R. Lewis Sr., John Wiley & Sons, Ltd, 1999, hard cover

Description

General description

Since its inception in the early 1950s, this work has become the bible for those who need to evaluate the hazard of substances used in commerce. It is the only reference that combines, for so many substances, data on toxicological, fire, reactivity, explosive potential, and regulatory information. Highly acclaimed in the professional journals, it was reviewed as an indispensable reference guide for professionals responsible for managing industrial safety and occupational exposure...an extraordinary work, and as a monumental work...Efforts like this are the things of what history is made, among other reviews.

Table of Contents

DOT Guide Number Cross-Index.
CAS Number Cross-Index.
Synonym Cross-Index.
References.
Volume II General Chemicals: Entries A - G.
Volume III General Chemicals: Entries H - Z.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?