• USA Home
  • Z507385 - Bouton Bold series safety spectacles
EMAIL THIS PAGE TO A FRIEND
Z507385 Aldrich
Bouton Bold series safety spectacles
Professional, clear Lens
Purchase
Properties
Related Categories Eye Protection Equipment, Eyeshields, Labware, Safety Equipment
mfr. no.   Bouton
availability   not available in EU
color   clear Lens
Description
General description
Polycarbonate lenses supported on black Resilium nylon frame, with a Soft-Fit nose bridge to prevent slippage. 100% UV protection. Meets ANSI Z87.1 standards.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?