EMAIL THIS PAGE TO A FRIEND
Z548235 Sigma-Aldrich

Wet wipes

L × W 10 in. × 12 in.

Purchase

Properties

Related Categories Labware, Wipers More...
packaging   pkg of 70 ea
L × W   10 in. × 12 in.

Description

General description

Contains citrus scented degreaser for dissolving grease and grime. Dual textured, industrial strength towel.

Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?