• USA Home
  • Z553476 - Industrial Dyes: Chemistry, Properties, Applications
EMAIL THIS PAGE TO A FRIEND
Z553476 Aldrich
Industrial Dyes: Chemistry, Properties, Applications
K. Hunger, John Wiley & Sons, 2003, 500 pp., hard cover
  •  ISBN-10 3527304266
  •  ISBN-13 978-352730426-4
Purchase
Properties
publication info   K. Hunger, John Wiley & Sons, 2003, 500 pp., hard cover
Description
General description
Dyes - originally obtained exclusively from natural sources - are today also produced synthetically on a large scale and represent one of the very mature and traditional sectors of the chemical industry. The present reference work on Industrial Dyes provides a comprehensive review of the chemistry, properties and applications of the most important groups of industrial dyes, including optical brighteners. It also outlines the latest developments in the area of functional dyes. Renowned experts in their respective fields have contributed to the chapters on chemical chromophores, synthesis and application of the various dye classes, textile dyeing and non-textile dyeing.
Table of Contents
Preface.
List of Contributors.
Dyes, General Survey.
Important Chemical Chromophores of Dye Classes.
Dye Classes for Principal Applications.
Textile Dyeing.
Nontextile Dyeing.
Functional Dyes.
Optical Brighteners.
Health and Safety Aspects.
List of Examples of Commercially Available Dyes.
Index.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?